Learn More
AGI-5198, MedChemExpress
Product Code. 16467458
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16467458
Brand: Medchem Express HY18082
MedChemExpress AGI-5198 (IDH-C35) is a potent and selective mutant IDH1R132H inhibitor with an IC50 of 0.07 μM.
AGI-5198 (IDH-C35) is a potent and selective mutant IDH1R132H inhibitor with an IC50 of 0.07 μM.
Specifications
Chemical Name or Material AGI-5198
CAS 1355326-35-0
Color White
Purity Grade Notes Research
Molecular Formula C27H31FN4O2
Quantity 10 mg
Synonym IDH-C35
Solubility Information DMF : ≥ 50 mg/mL (108.09 mM) ∣DMSO : 20.83 mg/mL (45.03 mM; Need ultrasonic)
SMILES FC1=CC=CC(N(C(C2=CC=CC=C2C)C(NC3CCCCC3)=O)C(CN4C=CN=C4C)=O)=C1
Molecular Weight (g/mol) 462.56
Formula Weight 462.56
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death