Learn More
Giese™ Agate Mortar with Pestle
Click to view available options
Capacity (Metric):
6 mL
8 mL
10 mL
13 mL
15 mL
24 mL
30 mL
50 mL
80 mL
130 mL
200 mL
350 mL
Unit Size:
Each
Description
- Polished agate construction
- For grinding and crushing
- Durable
- Available in different sizes
Specifications
Specifications
| Capacity (Metric) | 30 mL |
| Product Type | Mortar and Pestle |
| Material | Agate Stone |