Learn More
Thermo Scientific Chemicals Agate Mortar and Pestle, 95mm OD
Product Code. p-6684013
Click to view available options
Diameter (Metric) Outer:
65mm
95mm
Unit Size:
Each
Product Code. 15481557
Brand: Thermo Scientific Chemicals 039792.KT
Only null left
Add to basket
For preparing samples for the Quick Press
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Outer 3.7 in.
Diameter (Metric) Outer 95mm
Product Type Mortar and Pestle
For Use With (Equipment) Wig-L-Bug Mixing/Grinding Mill