Learn More
LNI Swissgas AG OFCAS 35
Product Code. 17882339 Shop All LNI Swissgas Products
Click to view available options
Type:
Compressor
Unit Size:
Each
Product Code. 17882339
Brand: LNI Swissgas 6920.90.035
Compressor with dryer integrated.
  • Compressor 35 L/min at 7 bar, 230 V plus with dryer integrated.
Specifications
Type Compressor