missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ag, Al, B, Ba, Bi, Ca, Cd, Cr, Cu, Fe, In, Li, K, Mg, Mn, Mo, Na, Ni, P, Pb, Sb, Si, Sn, Ti, V, Zn @ 900 μg/g in 75 cSt Hydrocarbon Oil, VHG™
Click to view available options
Quantity:
100 g
200 g
400 g
Unit Size:
100g
200g
400g
Description
Ag, Al, B, Ba, Bi, Ca, Cd, Cr, Cu, Fe, In, Li, K, Mg, Mn, Mo, Na, Ni, P, Pb, Sb, Si, Sn, Ti, V, Zn @ 900 μg/g in 75 cSt Hydrocarbon Oil
Specifications
Specifications
| Chemical Name or Material | Aluminium/Antimony/Barium/Bismuth/Boron/Cadmium/Calcium/Chromium/Copper/Indium/Iron/Lead/Lithium/Magnesium/Manganese/Molybdenum/Nickel/Phosphorus/Potassium/Silicon/Silver/Sodium/Tin/Titanium/Vanadium/Zinc |
| CAS | 7429-90-5, 7440-36-0, 7440-39-3, 7440-69-9, 7440-42-8, 7440-43-9 |
| Methods | ICP/XRF/RDE |
| Analyte or Component Names | Aluminium/Antimony/Barium/Bismuth/Boron/Cadmium/Calcium/Chromium/Copper/Indium/Iron/Lead/Lithium/Magnesium/Manganese/Molybdenum/Nickel/Phosphorus/Potassium/Silicon/Silver/Sodium/Tin/Titanium/Vanadium/Zinc |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Hydrocarbon Oil |
| Quantity | 200 g |
| Concentration or Composition (by Analyte or Components) | Aluminium 900 μg/g/Antimony 900 μg/g/Barium 900 μg/g/Bismuth 900 μg/g/Boron 900 μg/g/Cadmium 900 μg/g/Calcium 900 μg/g/Chromium 900 μg/g/Copper 900 μg/g/Indium 900 μg/g/Iron 900 μg/g/Lead 900 μg/g/Lithium 900 μg/g/Magnesium 900 μg/g/Manganese 900 μg/g/Molybdenum 900 μg/g/Nickel 900 μg/g/Phosphorus 900 μg/g/Potassium 900 μg/g/Silicon 900 μg/g/Silver 900 μg/g/Sodium 900 μg/g/Tin 900 μg/g/Titanium 900 μg/g/Vanadium 900 μg/g/Zinc 900 μg/g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction