missing translation for 'onlineSavingsMsg'
Learn More

AF 12198, TRC

Product Code. 17924265
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17924265 5 mg 5mg
17974605 1 mg 1mg
17982195 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17924265 Supplier Toronto Research Chemicals Supplier No. TRCA3550205MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 185413-30-3
Synonym N-Acetyl-L-phenylalanyl-L-α-glutamyl-L-tryptophyl-L-threonyl-L-prolylglycyl-L-tryptophyl-L-tyrosyl-L-glutaminyl-(2S)-2-azetidinecarbonyl-L-tyrosyl-L-alanyl-L-leucyl-L-prolyl-L-Leucinamide∣AF 12198

Specifications

Format Neat
Molecular Weight (g/mol) 1895.12
Quantity 5 mg
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.