Learn More
Eppendorf™ Aerosol-Tight Rotor Lids
Shop All Eppendorf Products
Click to view available options
For Use With (Equipment):
FA–45-24-11
FA–45-30-11
Rotor A-4-81
Unit Size:
Each
Pack of 2
Description
- Aluminim construction
- Aerosol-tight
- Autoclavable for 20 min at 121°C
Specifications
Specifications
| Type | Aerosol-tight Lid |
| Material | Aluminum |
| For Use With (Equipment) | FA–45-24-11 |
| Autoclavable | Autoclavable (121°C for 20 min.) |