Learn More
Thermo Scientific™ Rehydration Fluid for Culti-Loops Organisms, Aerobic Set
Click to view available options
Quantity:
10 Vials/Set
Unit Size:
Pack of 10
Description
Rehydration fluid set for Thermo Scientific™ Culti-Loops™ aerobic organisms.
Specifications
Specifications
| Description | Aerobic Set Rehydration Fluid |
| Format | Vial |
| Product Type | Quality Control |
| Target Organism Class | Organism Sets |
| Packaging Type | 10 Vial/Set |
| Quantity | 10 Vials/Set |
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.