missing translation for 'onlineSavingsMsg'
Learn More

AEE788, MedChemExpress

Product Code. 16485757
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16485757

Brand: Medchem Express HY10045

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AEE788 is an inhibitor of the EGFR and ErbB2 with IC50 values of 2 and 6 nM, respectively.

AEE788 is an inhibitor of the EGFR and ErbB2 with IC50 values of 2 and 6 nM, respectively.

Specifications

Chemical Name or Material AEE788
Physical Form Solid
CAS 497839-62-0
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C27H32N6
For Use With (Application) Cancer-Kinase/protease
Synonym NVP-AEE 788
Solubility Information DMSO : 50 mg/mL (113.49 mM; Need ultrasonic)
SMILES C[C@H](C1=CC=CC=C1)NC2=C(C=C3C(C=C4)=CC=C4CN5CCN(CC)CC5)C(N3)=NC=N2
Molecular Weight (g/mol) 440.58
Formula Weight 440.58
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.