Learn More
AEBP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11232-2-AP-20UL
This item is not returnable.
View return policy
Description
AEBP2 is a DNA-binding transcriptional repressor. It may interact with and stimulate the activity of the PRC2 complex, which methylates ′Lys-9′ and ′Lys-27′ residues of histone H3.Specifications
| AEBP2 | |
| Polyclonal | |
| Unconjugated | |
| AEBP2 | |
| AE binding protein 2, AEBP2, Zinc finger protein AEBP2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11569, 121536, 297705 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6ZN18, Q9Z248 | |
| Aebp2 | |
| AEBP2 Fusion Protein Ag1756 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |