missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Adrenomedullin Monoclonal Antibody (C1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543984
This item is not returnable.
View return policy
Description
Adrenomedullin (ADM), a vasodilator produced by most contractile cells, is characterized by persistent hypotensive activity. ADM is involved in the regulation of fluid and electrolyte homeostasis and in the maintenance of cardiovascular functioning. In hypertensive patients, the level of ADM in plasma is upregulated. Natriuresis is a common systemic manifestation of aneurysmal subarachnoid hemorrhage. ADM has strong natriuretic actions. ADM-induced natriuresis is caused by an increase in glomerular filtration rate and a decrease in distal tubular sodium reabsorption. ADM is present both in the periphery and brain, and can exert central effects such as decreasing food ingestion.
Specifications
| Adrenomedullin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P35318 | |
| ADM | |
| Ala22-Gly147 (Accession P35318), with N-terminal His Tag. | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| Western Blot | |
| C1 | |
| Unconjugated | |
| ADM | |
| ADM; Adrenomedullin; AM; AMpreproadrenomedullin; Ap; H39316; PAMP; preproadrenomedullin; proadrenomedullin N-20 terminal peptide; ProAM N-terminal 20 peptide; ProAM-N20; RATAP | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 133 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur