Learn More
MedChemExpress Adrenocorticotropic Hormone (ACTH) (4-10), human
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Purity | 99.49% |
| CAS | 4037-01-8 |
| Content And Storage | Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month |
| For Use With (Application) | Metabolism-protein/nucleotide metabolism |
| Molecular Weight (g/mol) | 962.09 g/mol |
| Product Type | Peptides |
| Quantity | 25 mg |
| Shipping Condition | Room temperature |
| Solubility | DMSO : 25 mg/mL (25.99 mM∣Need ultrasonic) |