missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Adrenocorticotropic Hormone (ACTH) (1-10), human

Product Code. 16474066
Encompass_Preferred
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 16474066

Brand: MedChemExpress HYP1518

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Adrenocorticotropic Hormone (ACTH) (1-10), human, an adrenocorticotropin hormone fragment, possesses a weak α-melanocyte stimulating hormone (α-MSH) potency only at high doses (100 and 1000 nM).

Specifications

Purity 98.53%
CAS 2791-05-1
Content And Storage Protect from light, stored under nitrogen∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light, stored under nitrogen)
For Use With (Application) Metabolism-protein/nucleotide metabolism
Molecular Weight (g/mol) 1299.41 g/mol
Product Type Peptides
Quantity 25 mg
Shipping Condition Room temperature
Solubility DMSO : 250 mg/mL (192.40 mM∣Need ultrasonic) ∣H2O : 1 mg/mL (0.77 mM∣Need ultrasonic)
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.