Learn More
ADRB3 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30713-1-AP-20UL
This item is not returnable.
View return policy
Description
The ADRB3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ADRB3. This antibody recognizes human antigen. The ADRB3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ADRB3 | |
| 1100 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| ADRB3, ADRB3R, adrenergic, beta 3 , receptor, B3AR, Beta 3 adrenergic receptor, Beta 3 adrenoceptor, Beta 3 adrenoreceptor, BETA3AR, FLJ99960 | |
| 20 μL | |
| 155 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |