Learn More
3M™ Adflo™ Powered Air Purifying Respirator System with Speedglas™ 9100 MP Welding Helmet
Product Code. 17895656
Click to view available options
:
Welding Helmet
Product Code. 17895656
Brand: 3M™ 7100054792_CLEAN
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The Adflo™ Powered Air Purifying Respirator System with 3M™ Speedglas™ 9100 MP Welding Helmet is designed to help protect the eyes and face from radiation, sparks, and spatter.
  • The flip-up functionality built into the 3M™ Speedglas™ 9100 MP Welding Helmet with 3M™ Adflo™ Powered Air Purifying Respirator System makes it an adaptable protection solution.
  • This system allows for quick swapping between welding filter and clear view, helping the user inspect their work while they go.
TRUSTED_SUSTAINABILITY
Specifications
Battery Life Turbo: approx. 7-9 hours || ADF: up to 1800 hours
Battery Type ADF: Lithium 3V || Turbo: Lithium-ion
Cartridge/Filter COMBINATION
Color Black
Filter Type High Efficiency (HE)
Product Type Auto-darkening Filter
Size One Size
Includes 9100-Air Welding Helmet, HE Filter, Lithium Ion Battery, Auto-Darkening Filter
Intrinsic Safety (IS) Rating No