Learn More
3M™ Adflo™ Heavy Duty Lithium Ion Battery
Product Code. 15710018
Click to view available options
:
Battery Type: Lithium ion
Product Code. 15710018
Brand: 3M™ 7000144539
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
3M™ Adflo™ Heavy Duty Lithium Ion Battery makes the Adflo™ Powered Air Respirator even more lightweight and balanced for greater comfort during the full shift.
  • Fully charges in 5.5 hours. Expected operating time is 10-13 hours with new particle filter, new fully-charged battery and a standard airflow
  • Batteries may be stored on the charger, fully charged, ready for immediate use
  • For use with the 3M™ Adflo™ Powered Air Respirator
TRUSTED_SUSTAINABILITY
Specifications
Battery Life Turbo: approx. 10 to 13 hr.
Battery Type Lithium-ion