missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Adenylate Cyclase 7 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17189183
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17189183 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17189183 Supplier Invitrogen™ Supplier No. PA5111269

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: PFAHLNHRES VSSGETHVPN GRRPKSVPQR HRRTPDRSMS PKGRSEDDSY DDEMLSAIEG LSSTRPCCSK SDDFYTFGSI FL Highest antigen sequence indentity to the following orthologs: Mouse - 79%, Rat - 79%.

This gene encodes a membrane-bound adenylate cyclase that catalyses the formation of cyclic AMP from ATP and is inhibitable by calcium. The product of this gene is a member of the adenylyl cyclase class-4/guanylyl cyclase enzyme family that is characterized by the presence of twelve membrane-spanning domains in its sequences.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adenylate Cyclase 7
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ADCY7
Gene Accession No. P51828
Gene Alias AA407758; AC7; ADCY7; adenylate cyclase 7; adenylate cyclase type 7; Adenylate cyclase type VII; adenylyl cyclase 7; adenylyl cyclase type VII; ATP pyrophosphate-lyase 7; FLJ36387; KIAA0037
Gene Symbols ADCY7
Host Species Rabbit
Immunogen Recombinant protein fragment of Human ADCY7 (Product #RP-98571)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.