missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Adenylate Cyclase 7 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16619844
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16619844 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16619844 Supplier Invitrogen™ Supplier No. PA5103390

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ADCY7.

This gene encodes a membrane-bound adenylate cyclase that catalyses the formation of cyclic AMP from ATP and is inhibitable by calcium. The product of this gene is a member of the adenylyl cyclase class-4/guanylyl cyclase enzyme family that is characterized by the presence of twelve membrane-spanning domains in its sequences.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adenylate Cyclase 7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ADCY7
Gene Accession No. P51828, P51829
Gene Alias AA407758; AC7; ADCY7; adenylate cyclase 7; adenylate cyclase type 7; Adenylate cyclase type VII; adenylyl cyclase 7; adenylyl cyclase type VII; ATP pyrophosphate-lyase 7; FLJ36387; KIAA0037
Gene Symbols ADCY7
Host Species Rabbit
Immunogen A synthesized peptide derived from human ADCY7(Accession P51828), corresponding to amino acid residues A751-F801.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113, 11513, 84420
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.