Learn More
BINDER™ Additional Universal Ventilation
Shop All BINDER Products
Click to view available options
For Use With (Equipment):
BINDER Chambers Series VD, VDL
Unit Size:
Each
Description
- 3/8 in. male thread connection or hose with 8 mm internal diameter
- The additional connection is made of stainless steel
Specifications
Specifications
| Description | Additional universal ventilation - Additional universal connection for ventilation with ambient air or inert gas. Flow limiter thread (RP in.), and connection with 3/8 in. male thread. |
| For Use With (Equipment) | BINDER Chambers Series VD, VDL |
| Type | Access Port |
| Material | Stainless Steel |
| Size | Universal |
| Voltage | 120/230 V |