Learn More
Memmert™ Additional Stainless Steel Grid, electro-polished, For If450Bw
Product Code. 16387277 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
Models 450 and 750 Ovens
Unit Size:
Each
Product Code. 16387277
Brand: Memmert™ IF450bwE20182
Only null left
Add to basket
Add to basket
Ideal for ramp operation with rapid temperature changes during incubation, breeding or storage.
  • For use with compressor-cooled incubator models 450 and 750
  • Grid pattern allows for airflow
  • Manufactured from electro-polished stainless steel
Specifications
For Use With (Equipment) Models 450 and 750 Ovens
Material Stainless Steel
Type Grid