missing translation for 'onlineSavingsMsg'
Learn More

ADDA 5 hydrochloride, MedChemExpress

Product Code. 16444116
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16444116 25 mg 25mg
16432658 5 mg 5mg
16420459 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16444116 Supplier Medchem Express Supplier No. HYU00448

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ADDA 5 hydrochloride is a partial non-competitive inhibitor of cytochrome c oxidase (CcO), with IC50s of 18.93 μM and 31.82 μM for purified CcO from human glioma and bovine heart, respectively.

ADDA 5 hydrochloride is a partial non-competitive inhibitor of cytochrome c oxidase (CcO), with IC50s of 18.93 μM and 31.82 μM for purified CcO from human glioma and bovine heart, respectively.

Specifications

Chemical Name or Material ADDA 5 hydrochloride
CAS 473268-46-1
Color White
Purity Grade Notes Research
Molecular Formula C24H36ClNO2
Quantity 25 mg
Solubility Information DMSO : 65 mg/mL (160.10 mM; Need ultrasonic) ∣H2O : 2 mg/mL (4.93 mM; ultrasonic and warming and heat to 60°C)
SMILES OC(COCCC1(C2)CC3CC2CC(C3)C1)CN4CC5=C(C=CC=C5)CC4.[H]Cl
Molecular Weight (g/mol) 406.0
Formula Weight 406.0
Percent Purity 98.06%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.