Learn More
Eppendorf™ Adapters For Fixed-Angle Round Bottom Rotor
Shop All Eppendorf Products
Click to view available options
Capacity:
15 to 18 mL
5 mL
7 to 15 mL
Unit Size:
Pack of 2
Description
- Adapters
- For Eppendorf™ Fixed Angle Round-Bottom Rotors
Specifications
Specifications
| Type | Adapter |
| Tube Type | Round Bottom |
| Capacity | 7 to 15 mL |
| For Use With (Equipment) | Fixed angle rotor |
| Bore Size (Metric) | 16 mm |
| No. of Places | 16 |
| No. per Pack | 2 Pack |