Learn More
Eppendorf™ Adapters For 100mL Rectangular Bucket
Product Code. p-8003411 Shop All Eppendorf Products
Click to view available options
Capacity:
1.2 to 5 mL
1.5 to 2 mL
15 mL
18 to 120 mL
18 to 30 mL
2.6 to 7 mL
3 to 15 mL
30 to 50 mL
50 mL
50 to 75 mL
7 to 18 mL
Unit Size:
Pack of 2
Product Code. 10266861
Brand: Eppendorf™ 5804750004
For use with Eppendorf Tubes
  • Adapters for rectangular buckets
  • For use with Rotor A-4-44
  • Available in a variety of sizes for a wide range of applications
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
Capacity 1.2 to 5 mL
For Use With (Equipment) 100mL Rectangular Bucket
Bore Size (Metric) 11 mm
No. of Places 56
For Use With (Application) Standard, Vacutainer, Falcon and micro test tubes
No. per Pack 2 Pack