Learn More
Eppendorf™ Adapters for Blood Tubes
Shop All Eppendorf Products
missing translation for 'orderingAttributeHoverText'
Capacity:
2.6 to 7 mL
missing translation for 'unitSize'
Pack of 2
Description
- For use with Rotor F-34-6-38 and Eppendorf Centrifuge 5804/5804 R
- For 3 round bottom tubes
Specifications
Specifications
| Type | Adapter |
| Tube Type | Round Bottom |
| Capacity | 2.6 to 7 mL |
| For Use With (Equipment) | Eppendorf™ Rotor F-34-6-38 |
| Bore Size (Metric) | 13 mm |
| No. of Places | 18 |
| No. per Pack | 2 Pack |