Learn More
HUBER Adapters
Click to view available options
Connection Type:
M16 x 1 male thread to M30 x 1.5 male thread
M24 x 1.5 female thread to M30 x 1.5 male thread
M24 x 1.5 female thread to R1/2 male thread
M24 x 1.5 male thread to M24 x 1.5 male thread
M30 x 1.5 female thread to 3/4 NPT male thread
M30 x 1.5 male thread to G3/4 male thread
M30 x 1.5 male thread to M30 x 1.5 male thread
M38 x 1.5 female thread to 1 NPT male thread
M38 x 1.5 female thread to M30 x 1.5 male thread
M38 x 1.5 female thread to R3/4 male thread
Unit Size:
Each
Specifications
Specifications
| Connection Type | M30 x 1.5 female thread to 3/4 NPT male thread |
| Quantity | 1 Ea. |
| Weight (Metric) | 0.2 kg |
| Type | Adapter |