Learn More
Eppendorf™ Adapter for Rotor F-35-6-30
Product Code. 10321254 Shop All Eppendorf Products
Click to view available options
Capacity:
1 x 100 to 125 mm Centriprep
1 x 15 mL Tube
1 x 2.6 to 7 mL Tube
1 x 30 mL Tube
1 x 50 mL Conical Tube
1 x 9 to 15 mL Tube
6 x 50 mL
Bore Size (Metric):
Large rotor bore
Small rotor bore
Unit Size:
Pack of 2
Pack of 6
Product Code. 10321254
Brand: Eppendorf™ 5427723005
Adapter for Rotors designed for use with Eppendorf Centrifuges
  • Rotor F-35-6-30 holds 6 x 15/50mL conical or 12 × Vacutainer™ tubes
TRUSTED_SUSTAINABILITY
Specifications
Type Large Rotor Bore Adapter
Tube Type Centriprep
Capacity 1 x 100 to 125 mm Centriprep
For Use With (Equipment) Rotor F-35-6-30
Bore Size (Metric) Large rotor bore
For Use With (Application) For Centriplus Centrifugal Filter Units
No. per Pack 6/Pk.