Learn More
Eppendorf™ Adapter for Rotor F-35-6-30
Shop All Eppendorf Products
Click to view available options
For Use With (Equipment):
for Rotor F-35-6-30, large rotor bore
for Rotor F-35-6-30, small rotor bore
Max. Tube Length:
65 to 89 mm
90 to 110 mm
Unit Size:
Pack of 2
Description
- For Rotor F-35-6-30
Specifications
Specifications
| Type | Adapter |
| Tube Type | Round Bottom |
| Max. Tube Length | 65 to 89 mm |
| For Use With (Equipment) | for Rotor F-35-6-30, small rotor bore |
| No. of Places | 12 |