Learn More
Eppendorf™ Adapter for 250 mL Rectangular Bucket in Rotor FA - 6 x 250
Shop All Eppendorf Products
Click to view available options
For Use With (Equipment):
1 x 50 mL Conical Tube
1 x 50 mL Oak Ridge Round Centrifuge Tube
1 x 50 mL Skirted Tubes
1 x 85 mL Oak Ridge Round Centrifuge Tube
3 x 30 mL Oak Ridge Round Centrifuge Tubes
4 x Eppendorf Tubes 5.0 mL and 15 mL conical tubes. (Universal adapters required for 5 mL tubes.)
7 x 10 mL Oak Ridge Round Centrifuge Tubes
7 x 16 mL Oak Ridge Round Centrifuge Tubes
7 x Blood Collection Round Tubes: D16, 5.5 to 12.0 mL
8 x Blood Collection Round Tubes: D13, 2.6 to 8.0 mL
9 x FACS Round Tubes, 5 mL
Unit Size:
Pack of 2
Description
- Adapts a wide range of centrifuge tubes to fit in Rotor FA - 6 x 250
- Set of 2 pieces
Specifications
Specifications
| For Use With (Equipment) | 7 x 10 mL Oak Ridge Round Centrifuge Tubes |
| Description | For 250 mL rectangular bucket in Rotor FA-6x250 |
| No. of Places | 7 |
| No. per Pack | Set of 2 pieces |