missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAMTS1 Monoclonal Antibody (OTI1B4), TrueMAB™, OriGene
Description
ADAMTS1 Monoclonal antibody specifically detects ADAMTS1 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | ADAMTS1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI1B4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | RRM1 |
| Gene Accession No. | P23921 |
| Gene Alias | a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 1; a disintegrin and metalloproteinase with thrombospondin motifs 1 (ADAMTS-1); a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 1; a disintegrin-like and metallopeptidse (reprolysin type) with thrombospondin type 1 motif, 1; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 1; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 1; ADAM metallopeptidase with thrombospondin type 1 motif, 1; ADAMs; ADAMTS; ADAM-TS 1; Adamts1; ADAM-TS1; ADAMTS-1; C3-C5; human metalloproteinase with thrombospondin type 1 motifs; KIAA1346; metalloendopeptidases; METH1; METH-1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?