missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13952-1-AP-150UL
This item is not returnable.
View return policy
Description
ADAM2 is a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. ADAM2 is a subunit of an integral sperm membrane glycoprotein called fertilin, which plays an important role in sperm-egg interactions.This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This member is a subunit of an integral sperm membrane glycoprotein called fertilin, which plays an important role in sperm-egg interactions.Specifications
| ADAM2 | |
| Polyclonal | |
| Unconjugated | |
| ADAM2 | |
| ADAM 2, ADAM metallopeptidase domain 2, ADAM2, Cancer/testis antigen 15, CRYN1, CRYN2, CT15, Fertilin subunit beta, FTNB, PH 30, PH 30b, PH30, PH30 beta | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11495, 2515 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q60718, Q99965 | |
| Adam2 | |
| ADAM2 Fusion Protein Ag5063 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction