Learn More
Socorex™ Acura™ Manual 825 Micropipette Triopack
Description
- Ergonomic shape.
- Lightweight.
- Uultra-soft plunger stroke.
- Smart and reliable volume adjustment:
- State-of-the-art micrometric setting is performed smoothly and precisely by turning push button.
- Finely machined click-stops and free rotating cap prevent any unwanted volume alteration.
- Digital display of set volume on the front of the instrument is easily visible during pipetting.
- Autoclavable.
Specifications
Specifications
| Certifications/Compliance | EN ISO 8655, DIN 12650, GMP, GLP, GBEA, NCCLS and CE IVD Directive 98/79 EEC on medical devices |
| Autoclavable | Autoclavable |
| Product Line | Acura |
| Type | Micropipette Triopack |
| Volume (Metric) | 10-100 μL, 100-1000 μL, 0.5-5.0 mL |
| Includes | 3 Acura™ micropipettes, manual, spike samples, Qualitips™ 100μL/1000μL/5mL |
| Quantity | 3/Pk. |