missing translation for 'onlineSavingsMsg'
Learn More

Justrite Activated Carbon Filter

Activated Carbon Filter for Lithium Battery Cabinets

Brand:  Justrite JCBCSMP000

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 17702388

  • 490.00 EUR / Each
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Top-mounted, activated carbon filter, with pre-filter
  • Easy filter replacement behind a hinged panel with key lock
Specifications

Specifications

Activated Carbon Filter
40 cm
Lithium Battery Cabinets
JCBCSMP000
Steel
51 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers