Learn More
Cytiva (formerly Pall Lab) Acrodisc™ Syringe Filters with Supor™ EKV Membrane Optimized for Scale Up
Product Code. 17184441
Change view
Click to view available options
:
0.2 μm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17184441 0.2 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17184441 Supplier Cytiva (formerly Pall Lab) Supplier No. 4902
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Syringe filters ideal for biological and general research.
  • Superior flow rates and higher throughputs than competitive devices
  • For research applications only
TRUSTED_SUSTAINABILITY
Specifications
Material (Membrane) PES
Pore Size 0.2 μm
Sterility Sterile