missing translation for 'onlineSavingsMsg'
Learn More

Acetyl Propionate, TRC

Product Code. 30322145
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30322145 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30322145 Supplier Toronto Research Chemicals Supplier No. TRCA16766850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetic Propionic Anhydride
CAS 13080-96-1
Molecular Formula C5 H8 O3
Quantity 50 mg
Synonym Propanoic acid, anhydride with acetic acid, Propionic acid, anhydride with AcOH (6CI,7CI), Propionic acid, anhydride with acetic acid (8CI), Acetic acid, anhydride with propionic acid (8CI), Acetic propionic anhydride, Acetyl propionate
InChI Formula InChI=1S/C5H8O3/c1-3-5(7)8-4(2)6/h3H2,1-2H3
SMILES CCC(=O)OC(=O)C
IUPAC Name acetyl propanoate
Molecular Weight (g/mol) 116.1152
Formula Weight 116.0473
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.