Learn More
Acetyl-L-homoserine lactone, TRC
Specifications
Specifications
| Chemical Name or Material | Acetyl-L-homoserine lactone |
| CAS | 51524-71-1 |
| Molecular Formula | C6H9NO3 |
| Quantity | 25 mg |
| InChI Formula | InChI=1S/C6H9NO3/c1-4(8)7-5-2-3-10-6(5)9/h5H,2-3H2,1H3,(H,7,8)/t5-/m0/s1 |
| SMILES | CC(=O)N[C@H]1CCOC1=O |
| IUPAC Name | N-[(3S)-2-oxooxolan-3-yl]acetamide |
| Molecular Weight (g/mol) | 143.14 |
| Formula Weight | 143.0582 |