missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone 99% Ph.Eur., Solveco
Click to view available options
Quantity:
12 L
25 L
Unit Size:
12 L
25 L
Description
Acetone of PA and pharma quality for laboratory use.
Specifications
Specifications
| Chemical Name or Material | 2-propanone |
| CAS | 67-64-1 |
| Boiling Point | 56°C |
| Quantity | 12 L |
| Packaging | 1 L HDPE Bottle |
| Concentration | 99.90% |
| Molecular Weight (g/mol) | 58.08 g/mol |
| Percent Purity | 99.90% |
| Grade | Reagent Ph. Eur. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction