Learn More
Acetone 2,4-Dinitrophenylhydrazone-3,5,6-d3, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Acetone 2,4-Dinitrophenylhydrazone-d3 |
| CAS | 259824-57-2 |
| Molecular Formula | C9 D3 H7 N4 O4 |
| Quantity | 0.01 g |
| Synonym | 2-Propanone, (4,6-dinitrophenyl-2,3,5-d3)hydrazone (9 CI), Acetone-2,4-dinitrophenyl-3,5,6-d3-hydrazone, Acetone 2,4-Dinitrophenylhydrazone-d3, Acetone-2,4-dinitrophenylhydrazone-D3 (phenyl-D3), Acetone 2,4-Dinitrophenylhydrazone-3,5,6-d3 |
| InChI Formula | InChI=1 S/C9H10N4O4/c1-6(2)10-11-8-4-3-7(12(14)15)5-9(8)13(16)17/h3-5,11 H,1-2H3/i3D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c(c1NN=C(C)C)[N+](=O)[O-])[N+](=O)[O-] |
| IUPAC Name | 2,3,5-trideuterio-4,6-dinitro-N-(propan-2-ylideneamino)aniline |
| Molecular Weight (g/mol) | 241.219 |
| Formula Weight | 241.089 g/mol |
| Show More |