Learn More
Acetic Acid, 1M (1N) Solution, Scharlab
Product Code. 11362655
Change view
Click to view available options
Quantity:
1 L
Packaging:
HDPE Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11362655 1 L HDPE Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11362655 Supplier Scharlab Supplier No. AC03651000
Ready-to-use volumetric solution of accurate and reliable concentration for titration. Acetic Acid, 1M (1N) Solution, Scharlab has a concentration that is exact to within the narrowest confidence interval for use as a reference solution in quantitative analysis.
  • Standard volumetric solution is ready to use
  • Certificate of Analysis details the factor, uncertainty level and methods used
  • Traceable to the SRM of NIST
  • Packaged in durable, chemically resistant HDPE containers that fit into most titrators
Specifications
CAS 64-19-7
Color Colorless
Appearance Clear
Molecular Formula C2H4O2
Linear Formula CH3OOH
MDL Number MFCD00036152
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
SMILES CC(O)=O
Concentration 1mol/L
IUPAC Name acetic acid
Molecular Weight (g/mol) 60.05
PubChem CID 176
ChEBI CHEBI:15366
Formula Weight 60.05g/mol
Grade Checked by means of potentiometric methods using a sodium hydroxide standard solution, that was also checked against Scharlab potassium hydrogen phthalate volumetric standard
Concentration or Composition (by Analyte or Components) 1mL = 0.060g
Density 1.01g/cm3
Quantity 1 L
Packaging HDPE Bottle
Physical Form Liquid
Traceability to NIST Yes
Chemical Name or Material Acetic Acid