Learn More
Acetate buffer solution, pH 4.7 EU Pharmacopoeia, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1000mL
Unit Size:
1000mL
Specifications
Specifications
| CAS | 7732-18-5 |
| pH Range | 4.7 |
| Product Type | Acetate buffer solution |
| Purity or Quality Grade | Eu. Ph. Reference |
| Quantity | 1000mL |