Learn More
Honeywell Fluka™ Acetate Buffer Solution Ph 4.6, Honeywell Fluka™
Product Code. 15623190 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500mL
1 L
6 x 500mL
Unit Size:
1 L
500mL
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15623190 1 L 1 L
15633190 500mL 500mL
15643190 6 x 500mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15623190 Supplier Honeywell Fluka™ Supplier No. 310481L
Request Bulk or Custom Format
Sodium acetate / acetic acid, for complexometry
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Specifications
CAS 126-96-5
Quantity 1 L