Learn More
Thermo Scientific™ Medifuge Benchtop Centrifuge Spacers
Product Code. 15510141 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
2 cm
3 cm
Color:
Green
Yellow
Unit Size:
8 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Color unitSize
15510141 2 cm Green 8 units
15520141 3 cm Yellow 8 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15510141 Supplier Thermo Scientific™ Supplier No. 75008817
Get accessories for Medifuge small centrifuge
TRUSTED_SUSTAINABILITY
Specifications
Type Benchtop Centrifuge Spacer
For Use With (Equipment) Medifuge Benchtop Centrifuge
Color Green
Capacity 13 mL
Description Spacer
Length (Metric) 2 cm
Includes Set of 8