Learn More
Fisherbrand™ AccuDuo Cartridge
Product Code. 15489729 Shop All Fisherbrand Products
Change view
Click to view available options
Type:
H Pack
P Pack
U Pack
U Pack (low TOC)
U Pack w/ 0.2μm PES Capsule Filter
For Use With (Equipment):
Accu20
Accu20, Accu500
Accu500
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type For Use With (Equipment) unitSize
15489729 U Pack Accu20, Accu500 Each
15459719 H Pack Accu500 Each
15449719 P Pack Accu20 Each
15499729 U Pack w/ 0.2μm PES Capsule Filter Accu20, Accu500 Each
15409739 U Pack (low TOC) Accu20, Accu500 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15489729
Only null left
Add to basket
Ajouter au panier
Ensures optimum quality of the final product water. Fisher Scientific™ AccuDuo Cartridges provide dual column filtration for Accu20, Accu100, and Accu500 water systems.
  • Includes two sets of dual-column purification cartridges
  • Cartridges work sequentially to ensure thorough removal of ions and organics in water
Spécification
For Use With (Equipment) Accu20, Accu500
Type U Pack
For Use With (Application) Ultrapure water for critical laboratory applications
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.