Learn More
IKA™ Accessory Stirring Shafts for SteadyStir™ and Overhead Stirrers
Product Code. p-167142 Shop All IKA Products
Click to view available options
Model:
R 1300
R 1302
R 1311
R 1342
R 1345
R 1352
R 1375
Unit Size:
Each
Product Code. 10568881
Brand: IKA™ 0000757700
Only null left
Add to basket
Add to basket
Generate axial flow in the vessel with these stirring shafts. IKA™ Accessory Stirring Shafts for SteadyStir™ and IKA™ Overhead Stirrers are designed for medium to high speeds.
Used at low to medium speeds.
† PTFE-coated
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) 2.8 in.
Diameter (English) Shaft 0.3 in.
Diameter (Metric) 70 mm
Diameter (Metric) Shaft 8 mm
Length (English) 21.7 in.
Length (Metric) 550 mm
Model R 1375
Speed 800 rpm max.
Type Paddle Stirrer
Material Stainless Steel (316L)