Learn More
Cytiva Accessories for SE 250/260 Mini-vertical Gel Units: Spacer
Product Code. 11755393 Shop All Cytiva Products
Change view
Click to view available options
Thickness (Metric):
0.07 cm
0.1 cm
0.15 cm
Unit Size:
Pack of 2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thickness (Metric) unitSize
11755393 0.07 cm Pack of 2
10585215 0.1 cm Pack of 2
10209354 0.15 cm Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11755393 Supplier Cytiva Supplier No. SE2119T2.75
Only null left
Specifications
For Use With (Equipment) SE 250, SE 260 Mini Format Vertical Gel Electrophoresis Unit
Type Spacer
Thickness (English) 0.02 in.
Thickness (Metric) 0.07 cm
Length (Metric) 8 cm