Learn More
Merck Accessories for Samplicity™ Filtration Systems
Shop All Merck Products
Click to view available options
Type:
Replacement Lid
Tube Set Assembly
Material:
Silicone
Thermoplastic Elastomer
Unit Size:
Each
Description
To be used with the Samplicity™ G2 Filtration System.
Powered by vacuum, the system takes minimal manual effort to filter 1 to 8 samples into standard HPLC vials in seconds, with higher recovery than syringe filters provide.
Specifications
Specifications
| Color | Clear |
| Description | Nonsterile lid with doors for attachment of Millex filters/funnels to HPLC tubes |
| For Use With (Equipment) | Samplicity G2 Filtration System with Millex Filters |
| Material | Thermoplastic Elastomer |
| For Use With (Application) | Analytical sample prep; High Performance Liquid Chromatography (HPLC) |
| Housing Material | PVDF |
| Quantity | 1 Ea. |
| Sufficient For | Up to 8 samples |
| Type | Replacement Lid |