Learn More
Merck Accessories for Samplicity™ Filtration Systems
Shop All Merck Products
Click to view available options
Type:
Replacement Lid
Tube Set Assembly
Material:
Silicone
Thermoplastic Elastomer
Unit Size:
Each
Description
To be used with the Samplicity™ G2 Filtration System.
Powered by vacuum, the system takes minimal manual effort to filter 1 to 8 samples into standard HPLC vials in seconds, with higher recovery than syringe filters provide.
Specifications
Specifications
| For Use With (Equipment) | Samplicity G2 Filtration System |
| Material | Silicone |
| Sterility | Non-sterile |
| For Use With (Application) | Analytical sample prep; High Performance Liquid Chromatography (HPLC) |
| No. per Pack | 1 Ea. |
| Quantity | 1 Ea. |
| Type | Tube Set Assembly |