Learn More
Thermo Scientific™ Accessories for RT2 Hotplate, RT2 Basic and Advanced Hotplate Stirrers
Product Code. 15288894 Shop All Thermo Scientific Products
Click to view available options
Type:
3 Prong Clamp (60 mm Grip)
C-5, Clamp Holder
Heating Bath
Support Rod
Temperature Probe PT100
Temperature Probe PT100, SN-8-4
Transparent Shield (PC)
Unit Size:
Each
Product Code. 15288894
Brand: Thermo Scientific™ 88880141
Only null left
Add to basket
Add to basket
Use these accessories to optimize the performance of your Thermo Scientific™ RT2 Hotplate, RT2 Basic and Advanced Hotplate Stirrers.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 500 mL
Diameter (Metric) 180 cm (outer), 179 cm (inner)
Type Heating Bath