Learn More
Thermo Scientific™ Accessories for MaxQ™ 8000 Shakers
Product Code. 10023982 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Stand for 1 Stackable Shaker
Stand for 2 Stackable Shakers
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
10023982 Stand for 1 Stackable Shaker Each
10424992 Stand for 2 Stackable Shakers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10023982 Supplier Thermo Scientific™ Supplier No. 194222
Only null left
Add to basket
Add to basket
Accommodate various applications and budgets with these flexible and convenient Thermo Scientific™ MaxQ™ 8000 Shaker Accessories.
Accommodate various applications and budgets with these flexible and convenient Thermo Scientific™ MaxQ™ 8000 Shaker Accessories. With exceptional simplicity and flexibility, these shaker accessories are the optimal choice for busy labs.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Stand for 1 Stackable Shaker
Depth (English) 35.5 in.
Depth (Metric) 89.7 cm
Height (English) 29.6 in.
Height (Metric) 75.2 cm
Width (English) 45.5 in.
Width (Metric) 115.6 cm
Weight (English) 158 lb.
For Use With (Equipment) MaxQ 8000