Learn More
Fisherbrand™ Accessories for Fisherbrand™ Heating/Cooling Mini Shaker
Product Code. p-6731003 Shop All Fisherbrand Products
Click to view available options
Holds:
12 x 15 mL Conical Centrifuge Tubes
20 x 1.5 mL Microcentrifuge Tubes
20 x 2 mL Microcentrifuge Tubes
5 x 50 mL Conical Centrifuge Tubes
9 x 21 mm Tubes
Unit Size:
Each
Product Code. 15524090
Only null left
Add to basket
Add to basket
Optional accessories to enhance the Fisher Scientific Heating/Cooling Mini Shaker
TRUSTED_SUSTAINABILITY
Specifications
Holds 9 x 21 mm Tubes
Product Type Tube Block
Description 9 x 21mm Block for 20mm diameter tube; Well: 21.7mm dia X .45mm depth