Learn More
SymLine™ System Accessories
Product Code. p-7236267
Click to view available options
Product Type:
Blind Plug
Cabinet Feed-through
Grounding Cable
Mounting Bracket
Color:
Black
Silver
Yellow
For Use With (Equipment):
32 mm Dia. Pipe
Cabinet (28 mm Borehole, 120 mm Wall Thickness)
Pipe System
SCAT SafetySystem Funnels
SCAT™ FlexTubes
Solvent System
Unit Size:
Each
Product Code. 17307429
Brand: SymLine™ 108270
Only null left
Add to basket
Add to basket
Dovetail your SymLine™ System to suit your specific needs! Here, you will find a lot of useful ancillaries.
  • SymLine™ offers many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables to allow you to customize your SymLine™ system
  • All accessories are electrically dissipative and approved for use in laboratories with explosion hazards
Specifications
Color Yellow
Length (Metric) 3 m
Includes Grounding Cable, Clamp, Clip
For Use With (Application) SymLine™ System Customization
For Use With (Equipment) SCAT™ FlexTubes
Product Type Grounding Cable